Learn More
Dynalene Heat Transfer Fluids DYNALENE HC-50 5 GALLONS
Supplier: Dynalene Heat Transfer Fluids HC50
Dynalene HC, is a Potassium Format/water solution that is non-toxic, non-flammable and non-hazardous. This fluid is engineered to provide high performance heat transfer at low temperatures with a freezing point below -57 F. The Dynalene HC is available to serve a wide range of applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.