Learn More
MSC Dynalon Labware Automatic Titrating Burette, 25 mL, Borosilicate Glass, DIN ISO 385
Catalog No. 5000527736
Encompass
This automatic titrating burette is designed for various laboratory applications. It features a self-zeroing mechanism and precise dispensing capabilities, making it suitable for accurate titrations. The burette is constructed from Schott graduated borosilicate glass, while the chemically resistant bottle is made of low-density polyethylene.
  • Self-zeroing for convenient operation
  • Measured to deliver precise volumes
  • Schott graduated borosilicate glass burette with blue ring graduations and Schellbach stripe for easy measurements
  • Exact dispensing of precise discharge jet or drop via the micro screw
  • Designed for left or right-hand operation
  • Includes a sturdy stand for support
  • Comes with a 1 liter low-density polyethylene (LDPE) chemically resistant bottle
  • Graduations comply with Category B (Tolerances according to DIN ISO 385)
Catalog No. 50-005-27736 Supplier MSC Supplier No. 35674308
This automatic titrating burette is designed for various laboratory applications. It features a self-zeroing mechanism and precise dispensing capabilities, making it suitable for accurate titrations. The burette is constructed from Schott graduated borosilicate glass, while the chemically resistant bottle is made of low-density polyethylene.
  • Self-zeroing for convenient operation
  • Measured to deliver precise volumes
  • Schott graduated borosilicate glass burette with blue ring graduations and Schellbach stripe for easy measurements
  • Exact dispensing of precise discharge jet or drop via the micro screw
  • Designed for left or right-hand operation
  • Includes a sturdy stand for support
  • Comes with a 1 liter low-density polyethylene (LDPE) chemically resistant bottle
  • Graduations comply with Category B (Tolerances according to DIN ISO 385)