Learn More
MSC Dynalon Labware Safety Wash Bottle, Polyethylene, 3 inch Diameter, Wide Mouth
Catalog No. 5000527311
Encompass
This polyethylene safety wash bottle is designed for laboratory use, featuring a translucent body, a wide mouth, and includes a closure for secure containment. It has a 3 inch diameter and a height of 165 mm, suitable for containing liquids within a 100 to 999 mL volume capacity range.
  • Constructed from polyethylene for durability and chemical resistance.
  • Translucent body allows for easy visual inspection of contents.
  • Wide mouth design facilitates convenient filling and cleaning.
  • Comes with a secure closure to prevent spills and contamination.
  • Includes a write-on surface for clear and easy labeling.
  • Not autoclavable, ensuring specific handling requirements.
Catalog No. 50-005-27311 Supplier MSC Supplier No. 65361552
This polyethylene safety wash bottle is designed for laboratory use, featuring a translucent body, a wide mouth, and includes a closure for secure containment. It has a 3 inch diameter and a height of 165 mm, suitable for containing liquids within a 100 to 999 mL volume capacity range.
  • Constructed from polyethylene for durability and chemical resistance.
  • Translucent body allows for easy visual inspection of contents.
  • Wide mouth design facilitates convenient filling and cleaning.
  • Comes with a secure closure to prevent spills and contamination.
  • Includes a write-on surface for clear and easy labeling.
  • Not autoclavable, ensuring specific handling requirements.