Learn More
MSC Dynalon Labware Vented Wash Bottle, Polyethylene, 500 mL, Wide-Mouth
Catalog No. 5000512470
Encompass
This vented wash bottle is made from polyethylene and features a patented DripLok vapor venting valve to prevent solvent drips. Designed with a wide mouth for easy filling, this 500 mL bottle is translucent, allowing for easy viewing of contents, and includes a closure. It is also a custom write-on bottle with multi-lingual markings.
  • Patented DripLok vapor venting valve prevents solvent drips.
  • Wide-mouth design allows for easy filling.
  • Translucent body enables easy viewing of contents.
  • Custom write-on bottle with multi-lingual markings.
  • Includes a closure.
  • Suitable for use with ethanol and methanol.
  • Not autoclavable.
Catalog No. 50-005-12470 Supplier MSC Supplier No. 35659796
This vented wash bottle is made from polyethylene and features a patented DripLok vapor venting valve to prevent solvent drips. Designed with a wide mouth for easy filling, this 500 mL bottle is translucent, allowing for easy viewing of contents, and includes a closure. It is also a custom write-on bottle with multi-lingual markings.
  • Patented DripLok vapor venting valve prevents solvent drips.
  • Wide-mouth design allows for easy filling.
  • Translucent body enables easy viewing of contents.
  • Custom write-on bottle with multi-lingual markings.
  • Includes a closure.
  • Suitable for use with ethanol and methanol.
  • Not autoclavable.