Learn More
MSC Dynalon Labware Wide Mouth Bottle, 100 mL, PVC, Square, Screw Cap
Catalog No. 5000511025
Encompass
This wide mouth bottle is constructed from clear PVC and features a square design with a 100 mL capacity. It is equipped with a leakproof screw cap that includes a pulp and vinyl lined bakelite liner, providing excellent chemical resistance and a functional temperature range of -20°C to 80°C. The bottle is also lightweight.
  • Leakproof screw closure
  • Clear as glass construction
  • Wide mouth design
  • Good chemical resistance
  • Lightweight
  • Temperature range from -20°C to 80°C
  • Screw closure includes a pulp and vinyl lined bakelite liner
Catalog No. 50-005-11025 Supplier MSC Supplier No. 65361222
This wide mouth bottle is constructed from clear PVC and features a square design with a 100 mL capacity. It is equipped with a leakproof screw cap that includes a pulp and vinyl lined bakelite liner, providing excellent chemical resistance and a functional temperature range of -20°C to 80°C. The bottle is also lightweight.
  • Leakproof screw closure
  • Clear as glass construction
  • Wide mouth design
  • Good chemical resistance
  • Lightweight
  • Temperature range from -20°C to 80°C
  • Screw closure includes a pulp and vinyl lined bakelite liner