Learn More
MSC Dynalon Labware Wide-Mouth Bottle, Polyethylene, Autoclavable, Graduated
Catalog No. 5000516052
Encompass
This wide-mouth bottle from Dynalon Labware is made of polyethylene, designed for various laboratory applications. It is translucent, autoclavable, and features graduations in both milliliters and fluid ounces for precise liquid measurement. Each bottle includes a closure for secure containment.
  • Translucent for easy viewing of contents
  • Autoclavable for sterilization and reuse
  • Includes closure for secure sealing
  • Graduated in milliliters and fluid ounces for accurate volume readings
  • Wide-mouth design for convenient filling and pouring
  • Made from polyethylene for durability
Catalog No. 50-005-16052 Supplier MSC Supplier No. 82782798
This wide-mouth bottle from Dynalon Labware is made of polyethylene, designed for various laboratory applications. It is translucent, autoclavable, and features graduations in both milliliters and fluid ounces for precise liquid measurement. Each bottle includes a closure for secure containment.
  • Translucent for easy viewing of contents
  • Autoclavable for sterilization and reuse
  • Includes closure for secure sealing
  • Graduated in milliliters and fluid ounces for accurate volume readings
  • Wide-mouth design for convenient filling and pouring
  • Made from polyethylene for durability