Learn More
MSC Dynalon Labware Wide-Mouth Bottle, Polyvinyl Chloride, 500 mL, Clear, with Closure
Catalog No. 5000535454
Encompass
This Dynalon Labware wide-mouth bottle is constructed from clear polyvinyl chloride, offering good chemical resistance and a lightweight design. It has a 500 mL capacity and features a leakproof screw closure, making it suitable for various laboratory applications. The bottle can withstand temperatures ranging from -20°C to 80°C, and its closure is lined with pulp and vinyl bakelite.
  • Leakproof screw closure.
  • Clear as glass construction.
  • Wide mouth design.
  • Good chemical resistance.
  • Lightweight.
  • Temperature range from -20°C to 80°C.
  • Screw closures are pulp and vinyl lined bakelite.
Catalog No. 50-005-35454 Supplier MSC Supplier No. 65361255
This Dynalon Labware wide-mouth bottle is constructed from clear polyvinyl chloride, offering good chemical resistance and a lightweight design. It has a 500 mL capacity and features a leakproof screw closure, making it suitable for various laboratory applications. The bottle can withstand temperatures ranging from -20°C to 80°C, and its closure is lined with pulp and vinyl bakelite.
  • Leakproof screw closure.
  • Clear as glass construction.
  • Wide mouth design.
  • Good chemical resistance.
  • Lightweight.
  • Temperature range from -20°C to 80°C.
  • Screw closures are pulp and vinyl lined bakelite.