Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DynaMag™-96 Side Magnet

Catalog No. 12331D
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-331-D 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-331-D Supplier Invitrogen™ Supplier No. 12331D
Only null left
Add to Cart
Add to Cart

Includes

One magnet

Designed for both automation platforms (e.g., Tecan) and manual workflows, but is compatible with PCR-strips and 96-well plates (200 μL), non-skirted and half-skirted.

Designed for both automation platforms (e.g., Tecan) and manual workflows, but is compatible with PCR-strips and 96-well plates (200 μL), non-skirted and half-skirted.

  • High wash volumes: DynaMag-96 Side Magnet
  • Low elution volumes: DynaMag-96 Bottom magnet
  • With extra column (13 total)
  • Enables sample mixing simply by shifting 96-well plate back and forth from right-most position to left-most position
  • Shifting draws beads back and forth through sample and allows mixing to be achieved without handling, promoting faster processing and less sample handing-induced variation
  • When used in combination with DynaMag-96 Bottom magnet on automation platform, two magnets allow for significant volume changes through workflow

Magnet is not compatible with PCR tubes.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Description Magnet
Content And Storage Contains one magnet. Store at room temperature.
For Use With (Equipment) DynaMag™
Shipping Condition Room Temperature
Product Line DynaMag
Quantity 1 Each
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.