Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SampleRack (for DynaMag™-5 magnet)

Catalog No. 12304D
Encompass_Preferred
Change view
Click to view available options
Quantity:
8 samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-304-D 8 samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-304-D Supplier Invitrogen™ Supplier No. 12304D
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Requires

Magnet base

Replace or supplement magnets SampleRacks when using the DynaMag-5 magnet or when processing more than eight samples at a time.

Replace or supplement magnets SampleRacks when using the DynaMag-5 magnet or when processing more than eight samples at a time. The SampleRack intended for use with the base of the DynaMag-5 magnet. SampleRack holds up to eight cell samples at a time, with numbered positions (1 to 8) for easy control of samples and a design that ensures good sample visibility when placed on the magnet base (not supplied). Flexible clips hold tubes in place and accommodate tubes from 11–16 mm in diameter, including most standard flow tubes and blood collection tubes (Vacutainer; tubes), with an optimal working volume of 1–5 mL.

SampleRack DynaMag-5 magnet should not be used with standard 15 mL tubes (use use DynaMag-15 magnet).

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
For Use With (Equipment) DynaMag™-5
Product Line DynaMag
Quantity 8 samples
Shipping Condition Room Temperature
Product Type Sample Rack
What are the Gauss levels for the DynaMag-2 and DynaMag-Spin Magnets?

The DynaMag-2 Magnet is 3,500-3,700 minimum Gauss level and the DynaMag-Spin is 3,000-3,500 minimum Gauss level.

What is the standard conversion of Gauss to Tesla units?

1 Tesla is equivalent to 10,000 Gauss. .

How can I clean my DynaMag magnet?

Spray and/or wipe the DynaMag magnet unit with one of the following cleaning agents:

- 70% (v/v) isopropyl alcohol
- 1% (v/v) sodium hypochlorite solution (bleach)
- 0.1 N HCl solution

Do not submerge the magnet unit in aqueous solutions and avoid prolonged exposure to water or aqueous solutions. Clean with a damp cloth and mild detergent when exposed to harsh solvents. Do not autoclave the magnet unit as heating the magnet to above 70 degrees C can destabilize the magnetic material, resulting in loss of magnetic strength.

What is the difference between the DynaMag-2 Magnet (Cat. No. 12321D) and DynaMag-Spin Magnet (Cat. No. 12320D)?

The DynaMag-2 Magnet can hold 16 standard 1.5 mL or 2 mL microcentrifuge tubes, while the DynaMag-Spin Magnet holds up to 6 standard 1.5 mL microcentrifuge tubes.

How can I clean my DynaMag magnet?

Spray and/or wipe the DynaMag magnet unit with one of the following cleaning agents:

- 70% (v/v) isopropyl alcohol
- 1% (v/v) sodium hypochlorite solution (bleach)
- 0.1 N HCl solution

Do not submerge the magnet unit in aqueous solutions and avoid prolonged exposure to water or aqueous solutions. Clean with a damp cloth and mild detergent when exposed to harsh solvents. Do not autoclave the magnet unit as heating the magnet to above 70 degrees C can destabilize the magnetic material, resulting in loss of magnetic strength.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.