Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamic Diagnostics Acrylic Blood Boot Stand

Catalog No. 22899085
Change view
Click to view available options
Color:
Transparent
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-899-085 Transparent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-899-085 Supplier Dynamic Diagnostics Supplier No. 400041
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Holds blood unit for better organization

  • Sturdy, yet flexible stand allows easy insertion of blood bags
  • Transparent blood boot allows for easy reading of labels
  • Flat boot bottom allows for easy organiztion of blood units
  • Curved design helps prevent tears

Order Info

Dimensions: 4L x 2.125W x 3.25 in. H (10.2 x 5.4 x 8.25cm)

Specifications

Material Acrylic
Color Transparent
Product Type Blood Boot Stand
For Use With (Application) For easy organization of Blood units
Length (English) 4 in.
Length (Metric) 101 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.