Learn More
Dynamic Diagnostics Cubitainer™ Spill Trap
Catalog No. 22899207
Change view
Click to view available options
Type:
Spill Trap
Catalog No. Type
22-899-207 Spill Trap
1 options
Catalog No. 22-899-207 Supplier Dynamic Diagnostics Supplier No. 300033
Helps contain spills when dispensing reagents
Safely contains spills to help prevent slippery lab floors.
  • Fits over container neck without hampering dispensing
  • Use alone or combine with a folded paper towel or spill pillow (sold separately) for maximum absorption and easy clean-up
  • Durable stainless-steel construction provides years of use and withstands lab chemicals
  • Stabilizes spigot for complete drainage of contents
Specifications
Material Stainless Steel
Type Spill Trap
For Use With (Application) Prevents slippery lab floors
For Use With (Equipment) Container neck