Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamic Diagnostics Desktop Reference Display

Catalog No. 22899227
Change view
Click to view available options
Product Type:
Desktop Reference Display Starter Set
Index Tab
Wall Mount Reference Display Starter Set
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-899-227 Desktop Reference Display Starter Set
22-899-228 Wall Mount Reference Display Starter Set
22-899-230 Index Tab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-899-227 Supplier Dynamic Diagnostics Supplier No. 400070
  • $330.00
    $235.65 / Each of 1

    Save $94.35

    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Each display unit supplied with ten wire-reinforced, double-sided polypropylene pockets.

Organizes and protects important documents

  • Keep frequently referenced documents near commonly used instruments or phones
  • Holds up to twenty 8.5 x 11 in. (20.3 × 27.9cm) pages
  • Clear plastic pockets protect documents from spills, smudges and dirt; one-piece steel-wire frames keep pocket rigid yet flexible enough to move documents in and out of units
  • Optional swing arm accessory holds up to two units off the work surface, yet allows easy access

Specifications

Product Type Desktop Reference Display Starter Set
Color Clear
Includes Ten wire Reinforced, double-sided Polypropylene pockets
For Use With (Application) For organizes and protects important documents
For Use With (Equipment) Desktop model
Device Holds 20 8.5 x 11 in. Pages
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.