Learn More
Dynamic Diagnostics Floor Scraper
Catalog No. 22899260
Change view
Click to view available options
Length (English):
48 in.
Catalog No. Length (English)
22-899-260 48 in.
1 options
Catalog No. 22-899-260 Supplier Dynamic Diagnostics Supplier No. 210060
  • $131.00
    $107.65 / Each of 1
    Save $23.35
    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Help keep your laboratory environment clear of paraffin and hard-to-remove debris
  • The razor sharp blade eliminates strenuous scraping on all hard surface floors; lightweight and ergonomically designed
  • aluminum construction
Specifications
Product Type Floor Scraper
Length (English) 48 in.
Material Aluminum
For Use With (Application) Razor sharp blade eliminate strenuous scraping, blunt blade for delicate, uneven floor
Includes Canvas blade guard