Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamic Diagnostics Hooktote™ Phlebotomy Trays

Catalog No. 22898365
Change view
Click to view available options
Color:
Blue
Pink
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-898-365 Blue
22-124-972 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-898-365 Supplier Dynamic Diagnostics Supplier No. 220069BL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Center channel and hook design let the user hang this tray over most rails or foot boards

  • Hooks fold away and lock into position when not in use
  • Made of food-grade resin
  • Available in pink or blue

Specifications

For Use With (Equipment) Rails and foot boards
Product Type Hooktote™ Phlebotomy Tray
Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.