Learn More
Dynamic Diagnostics Control Dropper Tops
Catalog No. 22899079
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
22-899-079 Plastic
1 options
Catalog No. 22-899-079 Supplier Dynamic Diagnostics Supplier No. 200030
Add to Cart
Edge
Add to Cart
Designed for 5mL and 10mL threaded control vials
  • Allow precise control over dispensing of drops
  • Fits Thermo Scientific MAS™ Controls
  • Prevents over-pouring and spills
  • Prevents contamination that may result with transfer pipets
  • Just squeeze the dropper tip lightly for easy dispensing
  • Convenient snap-on tip cap preserves the integrity of controls
Specifications
Material Plastic