Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamic Diagnostics Isolation Station

Catalog No. 22899061
Change view
Click to view available options
Material:
Powder-coated Steel
Steel
Product Type:
Over-door Hanger
PCW Isolation Station
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type
22-899-061 Steel Over-door Hanger
22-899-013 Powder-coated Steel PCW Isolation Station
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-899-061 Supplier Dynamic Diagnostics Supplier No. 300020
Add to Cart
Add to Cart

Includes

This item (300020) is just the Over-the-Door Hanger. (Does not include the Isolation Station.)

Provides easy access to Personal Protection Equipment (PPE) in one area

  • Over-the-Door Hanger for use with PCW Isolation Station 300019
  • Allows the Isolation Station to be placed instantly when precautions are required

Specifications

Product Type Over-door Hanger
Includes Hardware
For Use With (Application) Provides easy access to personal protection equipment in one area
Color White
For Use With (Equipment) PCW isolation station
Material Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.