Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamic Diagnostics Magnetic Pockets

Catalog No. 22899235
Change view
Click to view available options
Product Type:
Magnetic Pockets
Repositionable Magnetic Pocket
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-899-235 Repositionable Magnetic Pocket
22-899-236 Magnetic Pockets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-899-235 Supplier Dynamic Diagnostics Supplier No. 400078
Add to Cart
Add to Cart

Allow easy posting of documents on metal surfaces

  • For use with 8.5 × 11 in. (20.3 × 27.9cm) documents
  • Magnetic back with double-sided opening for quick document loading
  • Repositionable pockets allow for fast corner closure to hold document perfectly

Specifications

Product Type Repositionable Magnetic Pocket
Color Black
Material PVC
For Use With (Application) Allow easy posting of documents on Metal surfaces
Device Holds 8.5 x 11 in. Documents
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.