Learn More
Dynamic Diagnostics Maxi Scraper
Catalog No. 22899264
Change view
Click to view available options
Material:
Steel
Catalog No. Material
22-899-264 Steel
1 options
Catalog No. 22-899-264 Supplier Dynamic Diagnostics Supplier No. 210064
Add to Cart
Add to Cart
Help keep your laboratory environment clear of paraffin and hard-to-remove debris
  • Contains a sharp side for removing stubborn objects as well as a dull side for removing harder materials, such as paraffin or compound
  • The ergonomic design
Specifications
Product Type Maxi Scraper
Material Steel
For Use With (Application) Locking blade with a sharp side for removing stubborn object and a dull side for removing hard material
Includes Cover