Learn More
Dynamic Diagnostics Mini Scraper
Description
- Ideal for cleaning around microtomes with its contoured design for easy gripping
- Push button lock system allows for easy changing of the blade
Specifications
Specifications
| Material | Plastic |
| For Use With (Application) | Contoured designed makes scraper easy to hold for easy cleaning around microtome's |
| Includes | Push button lock system |
| Product Type | Mini Scraper |