Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynamic Diagnostics Mobile Phlebotomy Workstations Accessories

Catalog No. 22899032
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-899-032 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-899-032 Supplier Dynamic Diagnostics Supplier No. 300053
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Type Large Equipment Basket
Color White
For Use With (Application) Ideal for storing smaller items
For Use With (Equipment) Phlebotomy Workstations
Height (English) 8.6 in.
Height (Metric) 21.8 cm
Includes Bracket
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.