Learn More
Vernier™ Dynamics Cart and Track System with Long Track
SDPSCI_ED
Catalog No. S15980ND Shop All Vernier Software Products
Change view
Click to view available options
Type: cart and track system
Catalog No.
S15980ND Type: cart and track system
1 options
Catalog No. S15980ND Supplier Vernier™ Supplier No. DTSLONG
Add to Cart
Add to Cart
Includes
Dynamics Cart and Track System with Long Track
Provide students with the tools they need to explore kinematics, dynamics, momentum, and energy.
  • This versatile system can also be easily adapted to study optics, color, and diffraction.
  • Four mass trays make it easy to vary the total cart mass.
  • Super-elastic trigger button allows for new types of collisions.
  • An anti-roll peg ensures that the cart rolls on the track but not off the table.
  • Top tray allows for easy attachment of sensors and masses.
  • Two cart colors (teal and grey) make it easier to discuss collisions.
Warranty and Services
5-year limited warranty
Specifications
Product Type Cart and Track System
For Use With (Application) Educational use only