Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dynamin I Antibody (YA4184) | 97 kDa | 10 UL
SDP

Catalog No. 5000446132
Encompass_Preferred

Dynamin I Antibody (YA4184) is a mouse-derived, non-conjugated IgG2a monoclonal antibody targeting Dynamin I. It is for research use only and not intended for medical applications.

  • Host: Mouse
  • Clonality: Monoclonal antibody
  • Reactivity: Human
  • Applications: WB, IHC-P, ELISA
  • Molecular weight: 97 kDa
  • Conjugation: Non-conjugated
  • Isotype: IgG2a
  • Formulation: Supplied in PBS with 0.05% sodium azide
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.

Catalog No. 50-004-46132 Supplier Medchemexpress LLC Supplier No. HYP8448710UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dynamin I Antibody (YA4184) is a mouse-derived, non-conjugated IgG2a monoclonal antibody targeting Dynamin I. It is for research use only and not intended for medical applications.

  • Host: Mouse
  • Clonality: Monoclonal antibody
  • Reactivity: Human
  • Applications: WB, IHC-P, ELISA
  • Molecular weight: 97 kDa
  • Conjugation: Non-conjugated
  • Isotype: IgG2a
  • Formulation: Supplied in PBS with 0.05% sodium azide
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.