Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dynamin I Antibody (YA4184) | 97 kDa | 100 UL
SDP

Catalog No. 5000446397
Encompass_Preferred

The Dynamin I Antibody (YA4184) is a mouse-derived, non-conjugated IgG2a monoclonal antibody designed for research applications. It specifically targets human Dynamin I and is suitable for various immunological assays.

  • Mouse-derived IgG2a monoclonal antibody
  • Targets Dynamin I
  • Reacts with human samples
  • Predicted and observed molecular weight: 97 kDa
  • Affinity purified
  • Non-conjugated and unmodified
  • Formulated in PBS with 0.05% sodium azide
  • Suitable for WB, IHC-P, and ELISA applications
  • Store at -20°C for 1 year, avoid repeated freeze/thaw cycles

Catalog No. 50-004-46397 Supplier Medchemexpress LLC Supplier No. HYP84487100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Dynamin I Antibody (YA4184) is a mouse-derived, non-conjugated IgG2a monoclonal antibody designed for research applications. It specifically targets human Dynamin I and is suitable for various immunological assays.

  • Mouse-derived IgG2a monoclonal antibody
  • Targets Dynamin I
  • Reacts with human samples
  • Predicted and observed molecular weight: 97 kDa
  • Affinity purified
  • Non-conjugated and unmodified
  • Formulated in PBS with 0.05% sodium azide
  • Suitable for WB, IHC-P, and ELISA applications
  • Store at -20°C for 1 year, avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.