Learn More
Medchemexpress LLC Dynamin I antibody (YA4184) | 97 kDa | 50 UL
SDP
Catalog No. 5000449350
Encompass_Preferred
Dynamin I Antibody (YA4184) is a mouse-derived, non-conjugated IgG2a monoclonal antibody targeting Dynamin I. This antibody is suitable for research use only.
  • Catalyzes the hydrolysis of GTP and mediates vesicle scission
  • Participates in many forms of endocytosis, such as clathrin-mediated endocytosis or synaptic vesicle endocytosis as well as rapid endocytosis (RE)
  • Applications include Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), and Enzyme Linked Immunosorbent Assay (ELISA)
  • Predicted and observed band size of 97 kDa
Catalog No. 50-004-49350 Supplier Medchemexpress LLC Supplier No. HYP8448750UL
Dynamin I Antibody (YA4184) is a mouse-derived, non-conjugated IgG2a monoclonal antibody targeting Dynamin I. This antibody is suitable for research use only.
  • Catalyzes the hydrolysis of GTP and mediates vesicle scission
  • Participates in many forms of endocytosis, such as clathrin-mediated endocytosis or synaptic vesicle endocytosis as well as rapid endocytosis (RE)
  • Applications include Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), and Enzyme Linked Immunosorbent Assay (ELISA)
  • Predicted and observed band size of 97 kDa