Learn More
TARGETMOL CHEMICALS INC DYNAMIN IN-2 5MG
SDP
Catalog No. 502579367
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dynamin IN-2 (compound 43) is a Wiskostatin analogue. Dynamin IN-2 is a potent dynamin inhibitor (dynamin I GTPase IC50= 1.0 uM). Dynamin IN-2 blocks clathrin mediated endocytosis (CME) (IC50= 9.5 uM). purity: 98%
Catalog No. 50-257-9367 Supplier TARGETMOL CHEMICALS INC Supplier No. T643695MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dynamin IN-2 (compound 43) is a Wiskostatin analogue. Dynamin IN-2 is a potent dynamin inhibitor (dynamin I GTPase IC50= 1.0 uM). Dynamin IN-2 blocks clathrin mediated endocytosis (CME) (IC50= 9.5 uM). purity: 98%