Learn More
Apexbio Technology LLC Dynamin inhibitory peptide 251634-21-6 25mg
SDP
Catalog No. 5000093268
Encompass_Preferred
Dynamin inhibitory peptide (CAS 251634-21-6) is a peptide-based inhibitor targeting the GTPase dynamin It is designed to competitively block the interaction between dynamin and amphiphysin proteins thereby inhibiting dynamin-dependent internalization pathways Dynamin inhibitory peptide exerts its biological activity primarily by disrupting dynamin amphiphysin binding This mechanism effectively inhibits endocytic processes and reduces receptor internalization including the cellular uptake of GABA receptors in neuronal models Based on these pharmacological properties Dynamin inhibitory peptide holds research potential in the study of vesicle trafficking inhibition of endocytic pathways and modulation of receptor internalization
Catalog No. 50-000-93268 Supplier Apexbio Technology LLC Supplier No. A104625
Dynamin inhibitory peptide (CAS 251634-21-6) is a peptide-based inhibitor targeting the GTPase dynamin It is designed to competitively block the interaction between dynamin and amphiphysin proteins thereby inhibiting dynamin-dependent internalization pathways Dynamin inhibitory peptide exerts its biological activity primarily by disrupting dynamin amphiphysin binding This mechanism effectively inhibits endocytic processes and reduces receptor internalization including the cellular uptake of GABA receptors in neuronal models Based on these pharmacological properties Dynamin inhibitory peptide holds research potential in the study of vesicle trafficking inhibition of endocytic pathways and modulation of receptor internalization