Learn More
Medchemexpress LLC Dynamin inhibitory peptide | 251634-21-6 | 96.1% | 1121.25 | 5 MG
SDP
Catalog No. 5000423095
Encompass_Preferred
Dynamin inhibitory peptide competitively blocks the binding of dynamin to amphiphysin, thus preventing endocytosis. It also blocks the dopamine D3 effect on GABAA receptors.
  • Competitively blocks dynamin binding to amphiphysin, preventing endocytosis
  • Blocks dopamine D3 effect on GABAA receptors
  • Inhibits GABAAR endocytosis at 50 μM in vitro
Catalog No. 50-004-23095 Supplier Medchemexpress LLC Supplier No. HYP10835MG
Dynamin inhibitory peptide competitively blocks the binding of dynamin to amphiphysin, thus preventing endocytosis. It also blocks the dopamine D3 effect on GABAA receptors.
  • Competitively blocks dynamin binding to amphiphysin, preventing endocytosis
  • Blocks dopamine D3 effect on GABAA receptors
  • Inhibits GABAAR endocytosis at 50 μM in vitro