Learn More
Gibco™ Dynamis™ Medium
Description
Gibco Dynamis liquid medium is specifically designed to offer the highest batch and fed-batch culture performance and yield with recombinant CHO cells in a chemically defined environment. The chemically defined, protein-free, animal origin component-free formulation provides the power to achieve high titers, start process development faster and streamline or simplify transfer to manufacturing scale.
Power to achieve higher titers
Dynamis Medium is fortified to help maximize recombinant CHO cell growth and protein expression in suspension batch culture, offering 170% greater production than select competitive media with simple glucose supplementation by day 14. Cultures grown in Dynamis Medium extend to even higher titers demonstrating good cell viability out to day 21. This medium can also provide up to 60% greater fed-batch growth and production over glucose-only fed-batch culture for selected cell lines and applications when used with EfficientFeed C+ Supplement (Cat. No. A2503104).
Power to start process development faster
As a fortified medium, Dynamis provides the power to get to a high cell density over 10 million cells per mL faster than competitive products and sustains that cell density longer by just maintaining glucose levels. The viable cell growth achieved make it a good choice for scale-up if you have started with CD FortiCHO medium. When paired with EfficientFeed C+ Supplement (Cat. No. A2503104), it provides a fast and efficient way to develop a process.
When to Use Dynamis Medium
- The cell line you are using is a transfected CHO K1, GS CHO or CHO-S cell line
- Maximum batch culture cell densities and protein titers are needed
- You have time to adapt cells into a new medium in an effort to maximize titers
- Cell health at harvest is a priority for downstream processing
When to Consider Another Gibco Medium
Consider CD OptiCHO Medium (Cat. No. 12681011) when:
- You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S cell lines
- Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
- Your CHO cell line is 'finicky,' i.e. hard to grow or adapt
Consider CD CHO Medium (Cat. No. 10743029) when:
- Your GS CHO cells are recommended to be grown in CD CHO medium
- Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)
Consider CD DG44 Medium (Cat. No. 12610010) when:
- You use parental DG44 or DXB11 CHO cells (dhfr-)
Design Your Own Media Specific to Your Needs
Whether it's a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
ActiCHO is a trademark of Cytiva, BalanCD is a registered trademark of Irvine Scientific Sales Co., Cellvento is a registered trademark of Merck KGAA, EXCELL is a registered trademark of Millipore Sigma, HyCell is a registered trademark of Cytiva, and PowerCHO is a registered trademark of Lonza.
Specifications
Specifications
| Cell Line | CHO DG44, CHO-S, cGMP Banked, CHO-S, CHO-K1, GS-CHO, HEK293 |
| Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
| Form | Liquid |
| Product Type | AGT Medium |
| Product Line | Dynamis |
| Quantity | 10 L |
| Shipping Condition | Room Temperature |
| Format | Universal Bag(s) |
| Content And Storage | Liquid format medium. Store in the dark at 2°C to 8°C. |
| Culture Type | Mammalian Cell Culture |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.