Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynarex™ BZK Antiseptic Towelettes

Catalog No. 19081438 Shop All Dynarex Corporation Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-081438 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-081438 Supplier Dynarex™ Supplier No. 1303
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Effective alcohol-free cleansing

  • For effective cleansing
  • Alcohol Free
  • Will not sting
  • Ideal for perineal and maternity care, clean catch kit use
  • Contains Benzalkonium Chloride solution U.S.P.
  • 5 x 7 in.

Specifications

Product Type Antiseptic Towelette
Form Wipe
For Use With (Application) Ideal for perineal and maternity care, clean catch kit use
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.