Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynarex™ Disposable Towel Drape

Catalog No. 19310671 Shop All Dynarex Corporation Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Material:
Tissue/Poly/Tissue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-310-671 Tissue/Poly/Tissue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-310-671 Supplier Dynarex™ Supplier No. 4410
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in minor surgical or therapeutic procedures where a sterile field or drape is required

  • Poly-interlined, 3-ply construction (tissue/poly/tissue) provides absorbency while preventing possible moisture or bacteria migration
  • Drape eliminates all labor costs associated with cloth towels or drapes
  • Non-fenestrated
  • Sterile
  • Measures 45.7 × 66.04cm (18 × 26 in.)

Specifications

Material Tissue/Poly/Tissue
Disposable Single-use
Product Type Towel Drape
Sterility Sterile
Latex Free Yes
For Use With (Application) For use in minor surgical or therapeutic procedures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.