Learn More
Medchemexpress LLC HY-121802 5mg Medchemexpress, Dynarrestin CAS:2222768-84-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1218025MG
Encompass_Preferred
Medchemexpress, HY-121802 5mg Dynarrestin CAS:2222768-84-3 Dynarrestin is a aminothiazole inhibitor of cytoplasmic dyneins 1 and 2. Dynarrestin rapidly and reversibly inhibits dynein 1-driven microtubule gliding in vitro plus a range of dynein 1- and 2-dependent processes in cells without affecting ATP hydrolysis and interfering with ciliogenesis. Dynarrestin suppresses hedgehog (Hh)-dependent proliferation of neuronal precursors and tumor cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9915