Learn More
Abcam Dynasore, dynamin inhibitor, 10MG
Supplier:  Abcam AB12019210MG
Encompass_Preferred
MW 322.3 Da, Purity >99%. Cell-permeable dynamin inhibitor, that blocks endocytosis. Non-competitively inhibits dynamin1, dynamin2 and mitochondrial dynamin GTPase activity. Selective over other small GTPases.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0769361