Learn More
TARGETMOL CHEMICALS INC DYNASORE 25MG
SDP
Catalog No. 502581077
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dynasore (Dynamin Inhibitor I) a cell-permeable chemical can reversibly and non-competitivity inhibit GTPase activity of dynamin 1/2 (IC50 15 uM in a cell-free assay). It also inhibits the mitochondrial dynamin Drp1 with no effect against other small GTPase. purity: 99%
Catalog No. 50-258-1077 Supplier TARGETMOL CHEMICALS INC Supplier No. T184825MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dynasore (Dynamin Inhibitor I) a cell-permeable chemical can reversibly and non-competitivity inhibit GTPase activity of dynamin 1/2 (IC50 15 uM in a cell-free assay). It also inhibits the mitochondrial dynamin Drp1 with no effect against other small GTPase. purity: 99%