Learn More
AdipoGen Dynasore,Chemical. CAS: 304448-55-3
SDP
Supplier:  AdipoGen AGCR10045M005
Encompass_Preferred
Dynasore, AdipoGen Life Sciences. Chemical. CAS: 304448-55-3 (free base) 1202867-00-2 (x H2O). Formula: C18H14N2O4 . 0.5H2O. MW: 322.3 . 9.0. Dynamin-1, -2 (DNM1, DNM2) and Dynamin-related protein 1 (DRP1) inhibitor. Endocytosis inhibitor. Viral infection inhibitor. Destabilizes actin filaments. Inhibits entry of Trypanosoma into macrophages. Inhibitor of mitophagy. Inhibitor of hepatocyte lipophagy
Catalog No. 50-464-625