Learn More
Selleck Chemical LLC Dynasore 50mg 304448-55-3
SDP
Supplier:  Selleck Chemical LLC S804750MG
Encompass_Preferred
Dynasore is a cell-permeable, reversible non-competitive dynamin inhibitor of GTPase activity of dynamin 1/2, with IC50 of 15 ?M in a cell-free assay, also inhibits the mitochondrial dynamin Drp1, with no effect against other small GTPase. Dynasore suppresses mTORC1 activity and induces autophagy. Dynasore inhibits the entry of several viruses, including herpes simplex virus (HSV). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5607