Learn More
Aviva Systems Biology Corp DYNC1I1 Antibody - N-terminal region (ARP52047_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52047_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DYNC1I1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DYNC1I1. Alternate names: DNCI1, DNCIC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60673