Learn More
Aviva Systems Biology Corp DYNC1LI1 Antibody - C-terminal region (ARP65751_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65751_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DYNC1LI1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human DYNC1LI1. Alternate names: DNCLI1, FLJ10219ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-561