Learn More
Julabo USA Inc DYNEO DD-200F.A R290 EA 230V
SDP
Catalog No. NC2747062
Encompass_Preferred
Refrigerated/heating circulator, JULABO, DYNEO DD-200F Analog, 230V/60Hz, R290 Refrigerant; -20…+200C; 2kW heater. Cooling power: 200W at 20C; 170W at 10C; 150W at 0C; 10W at -10C; 20W at -20C. Adjustable flow pump 8-27L/min, 1.5-10 PSI. USB, analog communication and USB data-logging. External Pt100 probe port. Color TFT screen, programmable, multi-lingual. M16x1 pump fittings with 8 and 12mm barbs. 3-4L bath tank. 9.1 x 15.4 x 25.6 in, 57lbs
Catalog No. NC2747062 Supplier Julabo USA Inc Supplier No. 9021701.N1.33.A
Add to Cart
Add to Cart
Request a Quote
Refrigerated/heating circulator, JULABO, DYNEO DD-200F Analog, 230V/60Hz, R290 Refrigerant; -20…+200C; 2kW heater. Cooling power: 200W at 20C; 170W at 10C; 150W at 0C; 10W at -10C; 20W at -20C. Adjustable flow pump 8-27L/min, 1.5-10 PSI. USB, analog communication and USB data-logging. External Pt100 probe port. Color TFT screen, programmable, multi-lingual. M16x1 pump fittings with 8 and 12mm barbs. 3-4L bath tank. 9.1 x 15.4 x 25.6 in, 57lbs