Learn More
Julabo USA Inc DYNEO DD-200F R290 EA 115V
SDP
Catalog No. NC2747056
Encompass_Preferred
Refrigerated/heating circulator, JULABO, DYNEO DD-200F, 115V/60Hz, R290 Refrigerant; -20…+200C; 1kW heater. Cooling power: 200W at 20C; 170W at 10C; 150W at 0C; 100W at -10C; 20W at -20C. Adjustable flow pump 8-27L/min, 1.5-10 PSI. USB communication and USB data-logging. External Pt100 probe port. Color TFT screen, programmable, multi-lingual. M16x1 pump fittings with 8 and 12mm barbs. 3-4L bath tank. 9.1 x 15.4 x 25.6 in, 58lbs
Catalog No. NC2747056 Supplier Julabo USA Inc Supplier No. 9021701.N1.02
Add to Cart
Add to Cart
Request a Quote
Refrigerated/heating circulator, JULABO, DYNEO DD-200F, 115V/60Hz, R290 Refrigerant; -20…+200C; 1kW heater. Cooling power: 200W at 20C; 170W at 10C; 150W at 0C; 100W at -10C; 20W at -20C. Adjustable flow pump 8-27L/min, 1.5-10 PSI. USB communication and USB data-logging. External Pt100 probe port. Color TFT screen, programmable, multi-lingual. M16x1 pump fittings with 8 and 12mm barbs. 3-4L bath tank. 9.1 x 15.4 x 25.6 in, 58lbs