Learn More
Julabo USA Inc DYNEO DD-BC26.D heating circulator, 115V/60Hz, 20…200C. 1kW heater. 26L tank. CSA/UL
SDP
Catalog No. 5000693592
Encompass_Preferred
Heating circulator, JULABO, DYNEO DD-BC26.D, 115V/60Hz, 20…200C. 1kW heater. Adjustable centrifugal pump 8-27LPM, 1.5-11.6 PSI. USB-A data-logging; USB-B and RS232 external communication. Lemosa Pt100 port for external temperature control and monitoring. Color screen with click and scroll navigation. Programmable. M16x1 pump fittings with 8 and 12mm barbs. 6...7.5L bath tank. Bath tank opening: 10.2x13.8/7.9 in (WxL/D); Dimensions: 15.4 x 24.4 x 18.9 in. 41.2lbs. CSA/UL marked.
Catalog No. 50-006-93592 Supplier Julabo USA Inc Supplier No. 9021526.22.D
Heating circulator, JULABO, DYNEO DD-BC26.D, 115V/60Hz, 20…200C. 1kW heater. Adjustable centrifugal pump 8-27LPM, 1.5-11.6 PSI. USB-A data-logging; USB-B and RS232 external communication. Lemosa Pt100 port for external temperature control and monitoring. Color screen with click and scroll navigation. Programmable. M16x1 pump fittings with 8 and 12mm barbs. 6...7.5L bath tank. Bath tank opening: 10.2x13.8/7.9 in (WxL/D); Dimensions: 15.4 x 24.4 x 18.9 in. 41.2lbs. CSA/UL marked.