Learn More
Selleck Chemical LLC Dyngo-4a
SDP
Catalog No. 5000000853
Encompass_Preferred
Dyngo-4a is a potent dynamin inhibitor with IC50 of 0 38 M 1 1 M and 2 3 M for DynI (brain) DynI (rec) and DynII (rec) respectively
Catalog No. 50-000-00853 Supplier Selleck Chemical LLC Supplier No. S71631G
Dyngo-4a is a potent dynamin inhibitor with IC50 of 0 38 M 1 1 M and 2 3 M for DynI (brain) DynI (rec) and DynII (rec) respectively