Learn More
Selleck Chemical LLC Dyngo-4a 50mg 1256493-34-1
SDP
Catalog No. 501364473
Encompass_Preferred
Dyngo-4a is a potent dynamin inhibitor with IC50 of 0.38 ?M, 1.1??M, and 2.3 ?M for DynI (brain), DynI (rec), and DynII (rec), respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4473 Supplier Selleck Chemical LLC Supplier No. S716350MG
Dyngo-4a is a potent dynamin inhibitor with IC50 of 0.38 ?M, 1.1??M, and 2.3 ?M for DynI (brain), DynI (rec), and DynII (rec), respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.