Learn More
Aviva Systems Biology Corp DYNLL1 Antibody - middle region (ARP51261_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51261_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DYNLL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DYNLL1. Alternate names: DLC1, DLC8, DNCL1, DNCLC1, LC8, LC8a, MGC126137, MGC126138, PIN, hdlc1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60529