Learn More
Aviva Systems Biology Corp DYNLL2 Antibody - N-terminal region (ARP52514_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52514_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DYNLL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DYNLL2. Alternate names: DNCL1B, Dlc2, MGC17810, RSPH22ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60758