Learn More
Aviva Systems Biology Corp DYNLRB1 Antibody - N-terminal region (ARP65558_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65558_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DYNLRB1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human DYNLRB1. Alternate names: BITH, BLP, DNCL2A, DNLC2A, ROBLD1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-519