Learn More
Medchemexpress LLC Dynole 2-24 | 1416313-72-8 | 99.07% | C24H41N3 | 1 ML
SDP
Catalog No. 5000355741
Encompass_Preferred
Dynole 2-24 is an indole-based dynamin GTPase inhibitor with an IC50 of 0.56 μM for dynamin I. It is nontoxic and demonstrates increased potency against dynamin I and II in vitro and in cells (IC₅₀(CME)=1.9 μM). The product also exhibits 4.4-fold selectivity for dynamin I and is an active in-cell inhibitor of clathrin-mediated endocytosis (CME).
  • Indole-based dynamin GTPase inhibitor
  • Nontoxic
  • Increased potency against dynamin I and II in vitro and in cells
  • 4.4-fold selectivity for dynamin I
  • Active in-cell inhibitor of clathrin-mediated endocytosis (CME)
Catalog No. 50-003-55741 Supplier Medchemexpress LLC Supplier No. HY14508010MM/1ML
Dynole 2-24 is an indole-based dynamin GTPase inhibitor with an IC50 of 0.56 μM for dynamin I. It is nontoxic and demonstrates increased potency against dynamin I and II in vitro and in cells (IC₅₀(CME)=1.9 μM). The product also exhibits 4.4-fold selectivity for dynamin I and is an active in-cell inhibitor of clathrin-mediated endocytosis (CME).
  • Indole-based dynamin GTPase inhibitor
  • Nontoxic
  • Increased potency against dynamin I and II in vitro and in cells
  • 4.4-fold selectivity for dynamin I
  • Active in-cell inhibitor of clathrin-mediated endocytosis (CME)