Learn More
Medchemexpress LLC Dynole 2-24 | 1416313-72-8 | 99.1% | 371.60 g·mol⁻¹ | C24H41N3 | 5 MG
SDP
Catalog No. 5000221310
Encompass_Preferred
Dynole 2-24 is an indole-based dynamin GTPase inhibitor used as a research reagent to inhibit dynamin I and II and to block clathrin-mediated endocytosis in cells.
  • Potent inhibitor of dynamin I (IC50 = 0.56 μM).
  • Active in cells to inhibit clathrin-mediated endocytosis (IC50 ≈ 1.9 μM).
  • Shows selectivity for dynamin I over dynamin II (≈4.4-fold).
  • High purity suitable for biochemical and cellular assays.
  • Available in small research quantities as an oil for convenient handling.
Catalog No. 50-002-21310 Supplier Medchemexpress LLC Supplier No. HY1450805MG
Dynole 2-24 is an indole-based dynamin GTPase inhibitor used as a research reagent to inhibit dynamin I and II and to block clathrin-mediated endocytosis in cells.
  • Potent inhibitor of dynamin I (IC50 = 0.56 μM).
  • Active in cells to inhibit clathrin-mediated endocytosis (IC50 ≈ 1.9 μM).
  • Shows selectivity for dynamin I over dynamin II (≈4.4-fold).
  • High purity suitable for biochemical and cellular assays.
  • Available in small research quantities as an oil for convenient handling.