Learn More
Medchemexpress LLC Dynole 2-24 | 1416313-72-8 | 99.1% | C24H41N3 | 100 MG
SDP
Catalog No. 5000356691
Encompass_Preferred
Dynole 2-24 is an indole-based dynamin GTPase inhibitor that is non-toxic and demonstrates increased potency against dynamin I and II both in vitro and in cells. It shows selectivity for dynamin I and is an active in-cell inhibitor of clathrin-mediated endocytosis.
  • Indole-based dynamin GTPase inhibitor
  • IC50 of 0.56 μM for dynamin I
  • Non-toxic
  • Increased potency against dynamin I and II in vitro and in cells
  • 4.4-fold selectivity for dynamin I
  • Active in-cell inhibitor of clathrin-mediated endocytosis (CME)
Catalog No. 50-003-56691 Supplier Medchemexpress LLC Supplier No. HY145080100MG
Dynole 2-24 is an indole-based dynamin GTPase inhibitor that is non-toxic and demonstrates increased potency against dynamin I and II both in vitro and in cells. It shows selectivity for dynamin I and is an active in-cell inhibitor of clathrin-mediated endocytosis.
  • Indole-based dynamin GTPase inhibitor
  • IC50 of 0.56 μM for dynamin I
  • Non-toxic
  • Increased potency against dynamin I and II in vitro and in cells
  • 4.4-fold selectivity for dynamin I
  • Active in-cell inhibitor of clathrin-mediated endocytosis (CME)