Learn More
Medchemexpress LLC Dynole 2-24 | 1416313-72-8 | 99.1% | C24H41N3 | 25 MG
SDP
Catalog No. 5000357371
Encompass_Preferred
Dynole 2-24 is an indole-based dynamin GTPase inhibitor with an IC₅₀ of 0.56 μM for dynamin I. This compound is non-toxic and demonstrates increased potency against dynamin I and II both in vitro and within cells, with an IC₅₀ (CME) of 1.9 μM. It exhibits a 4.4-fold selectivity for dynamin I. Dynole 2-24 acts as an active in-cell inhibitor of clathrin-mediated endocytosis (CME).
  • Indole-based dynamin GTPase inhibitor.
  • IC₅₀ of 0.56 μM for dynamin I.
  • Nontoxic.
  • Increased potency against dynamin I and II in vitro and in cells (IC₅₀(CME)=1.9 μM).
  • Shows 4.4-fold selectivity for dynamin I.
  • Active in-cell inhibitor of clathrin-mediated endocytosis.
Catalog No. 50-003-57371 Supplier Medchemexpress LLC Supplier No. 25MGHY145080
Dynole 2-24 is an indole-based dynamin GTPase inhibitor with an IC₅₀ of 0.56 μM for dynamin I. This compound is non-toxic and demonstrates increased potency against dynamin I and II both in vitro and within cells, with an IC₅₀ (CME) of 1.9 μM. It exhibits a 4.4-fold selectivity for dynamin I. Dynole 2-24 acts as an active in-cell inhibitor of clathrin-mediated endocytosis (CME).
  • Indole-based dynamin GTPase inhibitor.
  • IC₅₀ of 0.56 μM for dynamin I.
  • Nontoxic.
  • Increased potency against dynamin I and II in vitro and in cells (IC₅₀(CME)=1.9 μM).
  • Shows 4.4-fold selectivity for dynamin I.
  • Active in-cell inhibitor of clathrin-mediated endocytosis.