Learn More
Cayman Chemical Dynole 34-2 10mg
SDP
Catalog No. 502947839
Encompass_Preferred
A dynamin inhibitor (IC50s = 6.9 and 14.2 µM for dynamin 1 and dynamin 2 GTPase activity, respectively); induces multinucleation, inhibits cytokinesis, and induces apoptosis in HeLa cells at 10 µM; inhibits proliferation of several cancer cell lines (GI50s = 5.8-15 µM)
Catalog No. 50-294-7839 Supplier Cayman Chemical Supplier No. 3407310 MG
A dynamin inhibitor (IC50s = 6.9 and 14.2 µM for dynamin 1 and dynamin 2 GTPase activity, respectively); induces multinucleation, inhibits cytokinesis, and induces apoptosis in HeLa cells at 10 µM; inhibits proliferation of several cancer cell lines (GI50s = 5.8-15 µM)